Gene Transcripts
- Gene Symbol
- Lyn
- Entrez Gene ID
- 17096
- Gene Name
- Yamaguchi sarcoma viral (v-yes-1) oncogene homolog
- Gene Aliases
- AA407514, Hck-2, p53Lyn, p56Lyn
- Chromosome Location
- Chr. 4: 3678121 - 3791612 on Build GRCm38
- UniGene ID
- Mm.317331
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001111096.1 |
NP_001104566.1
|
8, 9 |
1038 |
|
NM_010747.2 |
NP_034877.2
|
8, 9 |
975 |
|
GenBank mRNA |
AK028112.1 |
- |
8, 9 |
971 |
|
AK151914.1 |
- |
8, 9 |
1036 |
|
AK152199.1 |
- |
8, 9 |
1036 |
|
AK153038.1 |
- |
8, 9 |
972 |
|
AK170561.1 |
- |
8, 9 |
1036 |
|
BC003253.1 |
- |
8, 9 |
915 |
3500419 |
BC031547.1 |
- |
8, 9 |
1008 |
3589207 |
BC157998.1 |
- |
8, 9 |
807 |
9007404 |
M57696.1 |
- |
8, 9 |
874 |
|
M57697.1 |
- |
8, 9 |
811 |
|
M64608.1 |
- |
8, 9 |
1024 |
|
View Full Product Details