Gene Transcripts

Gene Symbol
Gm10375
Entrez Gene ID
100042342
Gene Name
predicted gene 10375
Gene Aliases
ENSMUSG00000072602
Chromosome Location
Chr. 14: 43602627 - 43608013 on Build GRCm38
UniGene ID
Mm.429750
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098269.2 NP_001091739.1 5 549
GenBank mRNA BB679674.1 - 2 123
BC089536.1 - 5 549 6742411

Gene Symbol
1700001F09Rik
Entrez Gene ID
71826
Gene Name
RIKEN cDNA 1700001F09 gene
Gene Aliases
Gm10377
Chromosome Location
Chr. 14: 43342384 - 43347811 on Build GRCm38
UniGene ID
Mm.335855
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027940.2 NP_082216.1 5 597
GenBank mRNA AK005587.1 - 5 598
BC061060.1 - 5 579 6770724

Gene Symbol
Gm10377
Entrez Gene ID
100042055
Gene Name
predicted gene 10377
Gene Aliases
ENSMUSG00000072606
Chromosome Location
Chr. 14: 42792568 - 42797994 on Build GRCm38
UniGene ID
Mm.429109
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001244671.1 NP_001231600.1 5 560
GenBank mRNA BC049548.1 - 5 560

View Full Product Details