Gene Transcripts

Gene Symbol
Osbpl9
Entrez Gene ID
298369
Gene Name
oxysterol binding protein-like 9
Gene Aliases
Chromosome Location
Chr. 5: 128694407 - 128839435 on Build Rnor_6.0
UniGene ID
Rn.61554
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044234.2 NP_001037699.2 22 1981
GenBank mRNA BC105828.1 - 3 378
FQ228495.1 - 23 2037

View Full Product Details