Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013943.2 | NP_039234.1 | 4 | 672 | |
GenBank mRNA | AF097330.1 | - | 4 | 527 | |
AF109196.1 | - | 4 | 465 | ||
AK001624.1 | - | 4 | 559 | ||
AK095959.1 | - | 4 | 493 | ||
AK301466.1 | - | 4 | 470 | ||
AK307142.1 | - | 2 | 237 | ||
AK315051.1 | - | 4 | 509 | ||
AL117424.1 | - | 4 | 584 | ||
BC012444.1 | - | 4 | 537 | 3861372 | |
CR533501.1 | - | 4 | 387 |