Gene Transcripts

Gene Symbol
GNL3
Entrez Gene ID
26354
Gene Name
G protein nucleolar 3
Gene Aliases
C77032, E2IG3, NNP47, NS
Chromosome Location
Chr. 3: 52685920 - 52694497 on Build GRCh38
UniGene ID
Hs.313544
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014366.4 NP_055181.3 8, 9 948
NM_206825.1 NP_996561.1 8, 9 1045
NM_206826.1 NP_996562.1 8, 9 898
GenBank mRNA AF191018.1 - 8, 9 945
AK027514.1 - 8, 9 949
AK027516.1 - 8, 9 798
AK315484.1 - 8, 9 861
AY825265.1 - 7, 8 775
BC001024.2 - 8, 9 796 3342771

View Full Product Details