Gene Transcripts

Gene Symbol
OR2W1
Entrez Gene ID
26692
Gene Name
olfactory receptor family 2 subfamily W member 1
Gene Aliases
hs6M1-15
Chromosome Location
Chr. 6: 29044213 - 29045175 on Build GRCh38
UniGene ID
Hs.553526
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030903.3 NP_112165.1 1 644
GenBank mRNA BC103870.1 - 1 644
BC103871.1 - 1 644 40004106
BC103872.1 - 1 644 40004109

View Full Product Details