Gene Transcripts

Gene Symbol
CD274
Entrez Gene ID
29126
Gene Name
CD274 molecule
Gene Aliases
B7-H, B7H1, PD-L1, PDCD1L1, PDCD1LG1, PDL1
Chromosome Location
Chr. 9: 5450503 - 5470567 on Build GRCh38
UniGene ID
Hs.521989
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267706.1 NP_001254635.1 4 487
NM_014143.3 NP_054862.1 5 829
GenBank mRNA AF177937.1 - 4 721
AF233516.1 - 5 773
AK001894.1 - 3 536
AK300470.1 - 6 684
AK314567.1 - 5 790
AY254342.1 - 4 721
AY291313.1 - 3 379
BC069381.1 - 5 771 7262208
BC074984.2 - 4 741 30915301
BC113734.1 - 4 742 8322786
BC113736.1 - 4 742 8322788
DQ286582.1 - 3 379

View Full Product Details