Gene Transcripts

Gene Symbol
ZNF117
Entrez Gene ID
51351
Gene Name
zinc finger protein 117
Gene Aliases
H-plk, HPF9
Chromosome Location
Chr. 7: 64974452 - 64991036 on Build GRCh38
UniGene ID
Hs.250693
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015852.3 NP_056936.2 4 1391
GenBank mRNA BC034021.1 - 5 544
M27879.1 - 3 212
M55422.1 - 4 1390
M61867.1 - 3 212

View Full Product Details