Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001261.3 | NP_001252.1 | 1 | 174 | |
XM_017014184.1 | XP_016869673.1 | 1 | 40 | ||
GenBank mRNA | AB451289.1 | - | 1 | 51 | |
AB451421.1 | - | 1 | 51 | ||
AK313828.1 | - | 1 | 103 | ||
BC001968.1 | - | 1 | 111 | 3460767 | |
BT019903.1 | - | 1 | 51 | ||
DB120461.1 | - | 1 | 174 | ||
L25676.1 | - | 1 | 115 | ||
X80230.1 | - | 1 | 149 |