Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017420.4 | NP_059116.3 | 2 | 1186 | |
XM_005267759.2 | XP_005267816.1 | 3 | 1165 | ||
GenBank mRNA | AK308528.1 | - | 3 | 543 | |
AK309358.1 | - | 3 | 1165 | ||
BC098135.1 | - | 2 | 1063 | 40007527 | |
BC098282.1 | - | 2 | 1063 | 40007529 | |
BC099722.1 | - | 2 | 1063 | 40007530 | |
BC101934.1 | - | 2 | 1063 | 40007526 |