Gene Transcripts

Gene Symbol
CDX4
Entrez Gene ID
1046
Gene Name
caudal type homeobox 4
Gene Aliases
Chromosome Location
Chr. X: 73447254 - 73454585 on Build GRCh38
UniGene ID
Hs.553488
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005193.1 NP_005184.1 1 299
GenBank mRNA BC128233.1 - 1 299

View Full Product Details