Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020208.3 | NP_064593.1 | 7 | 1085 | |
NM_022405.3 | NP_071800.1 | 6 | 974 | ||
XM_011533847.2 | XP_011532149.1 | 6 | 1399 | ||
GenBank mRNA | AF125107.1 | - | 8 | 1149 | |
AJ276207.1 | - | 7 | 1012 | ||
AJ276208.1 | - | 6 | 901 | ||
AL389979.1 | - | 2 | 1149 | ||
BC126197.1 | - | 7 | 1025 | 8991913 | |
BC136431.1 | - | 7 | 1025 | 9020420 | |
BC144087.1 | - | 7 | 1025 | 9052607 |