Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304725.1 | NP_001291654.1 | 4 | 572 | |
NM_017655.5 | NP_060125.4 | 4 | 869 | ||
GenBank mRNA | AB073737.1 | - | 4 | 685 | |
AK000082.1 | - | 3 | 428 | ||
AK225941.1 | - | 5 | 708 | ||
AK307422.1 | - | 4 | 572 | ||
BC036075.1 | - | 5 | 869 | 5296057 |