Gene Transcripts
- Gene Symbol
- CLPTM1
- Entrez Gene ID
- 1209
- Gene Name
- CLPTM1, transmembrane protein
- Gene Aliases
- Chromosome Location
- Chr. 19: 44954585 - 44993346 on Build GRCh38
- UniGene ID
- Hs.444441
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282175.1 |
NP_001269104.1
|
5, 6 |
613 |
|
NM_001282176.1 |
NP_001269105.1
|
5, 6 |
637 |
|
NM_001294.3 |
NP_001285.1
|
5, 6 |
764 |
|
GenBank mRNA |
AF037339.1 |
- |
5, 6 |
607 |
|
AK074935.1 |
- |
5, 6 |
608 |
|
AK075262.1 |
- |
5, 6 |
607 |
|
AK223553.1 |
- |
5, 6 |
608 |
|
AK293314.1 |
- |
5, 6 |
629 |
|
AK304480.1 |
- |
5, 6 |
596 |
|
AK310826.1 |
- |
5, 6 |
637 |
|
AK316094.1 |
- |
5, 6 |
633 |
|
BC004865.2 |
- |
5, 6 |
643 |
3840991 |
BC012359.1 |
- |
5, 6 |
597 |
4685026 |
BI916245.1 |
- |
5, 6 |
610 |
|
BT007262.1 |
- |
5, 6 |
592 |
|
KU177986.1 |
- |
5, 6 |
592 |
|
KU177987.1 |
- |
5, 6 |
592 |
|
View Full Product Details