Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021035.2 | NP_066363.1 | 8 | 2717 | |
XM_006723836.1 | XP_006723899.1 | 8 | 2547 | ||
XM_011528924.1 | XP_011527226.1 | 7 | 2592 | ||
XM_017027971.1 | XP_016883460.1 | 6 | 1643 | ||
GenBank mRNA | AB037825.2 | - | 8 | 2535 | |
AK002139.1 | - | 8 | 2630 | ||
AK022641.1 | - | 6 | 1882 | ||
AK096594.1 | - | 8 | 2924 | ||
AK307392.1 | - | 8 | 2921 | ||
BC014041.2 | - | 7 | 2533 |