Gene Transcripts

Gene Symbol
CRYGB
Entrez Gene ID
1419
Gene Name
crystallin gamma B
Gene Aliases
CRYG2, CTRCT39
Chromosome Location
Chr. 2: 208142573 - 208146327 on Build GRCh38
UniGene ID
Hs.248102
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005210.3 NP_005201.2 3 317
XM_017003402.1 XP_016858891.1 3 497
GenBank mRNA BC074944.2 - 3 317
BC074945.2 - 3 317 30915496
BC117384.1 - 3 317 40125935
BC117388.1 - 3 317 40125939
BY794984.2 - 3 332
KT326704.1 - 3 322

View Full Product Details