Gene Transcripts

Gene Symbol
WDR25
Entrez Gene ID
79446
Gene Name
WD repeat domain 25
Gene Aliases
C14orf67
Chromosome Location
Chr. 14: 100376410 - 100530303 on Build GRCh38
UniGene ID
Hs.497600
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161476.1 NP_001154948.1 3 1049
NM_024515.4 NP_078791.3 3 1180
XM_005268056.4 XP_005268113.1 3 1056
XM_005268057.3 XP_005268114.1 2 206
XM_005268058.4 XP_005268115.1 2 220
XM_011537150.2 XP_011535452.1 3 1303
XM_011537151.2 XP_011535453.2 2 659
XM_017021639.1 XP_016877128.1 2 208
GenBank mRNA AK291960.1 - 3 1012
AY295082.1 - 3 1063
BC003641.2 - 2 286 3529568
CT003969.1 - 2 212

View Full Product Details