Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001308207.1 | NP_001295136.1 | 7 | 2752 | |
NM_001308208.1 | NP_001295137.1 | 6 | 2405 | ||
NM_025251.2 | NP_079527.1 | 5 | 2278 | ||
XM_011517308.1 | XP_011515610.1 | 6 | 2559 | ||
XM_011517309.1 | XP_011515611.1 | 6 | 2463 | ||
XM_011517312.2 | XP_011515614.1 | 4 | 3097 | ||
XM_017013870.1 | XP_016869359.1 | 7 | 3631 | ||
XM_017013871.1 | XP_016869360.1 | 6 | 2690 | ||
GenBank mRNA | AB051475.1 | - | 7 | 2756 | |
AK296353.1 | - | 6 | 2402 | ||
AK303851.1 | - | 2 | 981 |