Gene Transcripts

Gene Symbol
ATP13A4
Entrez Gene ID
84239
Gene Name
ATPase 13A4
Gene Aliases
Chromosome Location
Chr. 3: 193398967 - 193581725 on Build GRCh38
UniGene ID
Hs.674423
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032279.3 NP_115655.2 17 2250
XM_011513232.2 XP_011511534.1 17 1997
XM_017007318.1 XP_016862807.1 17 1886
XM_017007319.1 XP_016862808.1 17 1997
GenBank mRNA AK095277.1 - 17 2035
AK096510.1 - 8 846
AY823162.1 - 17 1927
BC101496.1 - 17 2021 8069002

View Full Product Details