Gene Transcripts

Gene Symbol
UMODL1
Entrez Gene ID
89766
Gene Name
uromodulin like 1
Gene Aliases
Chromosome Location
Chr. 21: 42062959 - 42142996 on Build GRCh38
UniGene ID
Hs.242520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004416.2 NP_001004416.2 11 1866
NM_001199527.1 NP_001186456.1 11 2046
NM_001199528.2 NP_001186457.2 11 2046
NM_173568.3 NP_775839.3 11 1866
XM_011529797.1 XP_011528099.1 9 1474
XM_017028506.1 XP_016883995.1 11 1861
XM_017028507.1 XP_016883996.1 11 4790
GenBank mRNA AB051810.1 - 11 2046
AB051811.1 - 11 2046
AY771619.1 - 11 1866
AY771620.1 - 11 1866

View Full Product Details