Gene Transcripts
- Gene Symbol
- CCDC149
- Entrez Gene ID
- 91050
- Gene Name
- coiled-coil domain containing 149
- Gene Aliases
- Chromosome Location
- Chr. 4: 24806117 - 24980204 on Build GRCh38
- UniGene ID
- Hs.106432
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130726.2 |
NP_001124198.1
|
6, 7 |
770 |
|
NM_173463.4 |
NP_775734.2
|
7, 8 |
871 |
|
XM_005248210.3 |
XP_005248267.1
|
6, 7 |
799 |
|
XM_011513906.2 |
XP_011512208.1
|
8, 9 |
881 |
|
XM_011513907.2 |
XP_011512209.1
|
7, 8 |
754 |
|
XM_011513908.1 |
XP_011512210.1
|
6, 7 |
676 |
|
XM_017008827.1 |
XP_016864316.1
|
8, 9 |
881 |
|
XM_017008828.1 |
XP_016864317.1
|
7, 8 |
805 |
|
GenBank mRNA |
AK125353.1 |
- |
6, 7 |
679 |
|
AK296454.1 |
- |
3, 4 |
428 |
|
AK302905.1 |
- |
7, 8 |
815 |
|
AL834257.2 |
- |
6, 7 |
744 |
|
DR423861.1 |
- |
6, 7 |
685 |
|
View Full Product Details