Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031909.2 | NP_114115.2 | 2 | 1006 | |
XM_017017165.1 | XP_016872654.1 | 3 | 2324 | ||
XM_017017166.1 | XP_016872655.1 | 3 | 2100 | ||
GenBank mRNA | AF329838.1 | - | 2 | 963 | |
BC035628.1 | - | 2 | 789 | 5199881 |