Gene Transcripts

Gene Symbol
PSMA8
Entrez Gene ID
143471
Gene Name
proteasome subunit alpha 8
Gene Aliases
PSMA7L
Chromosome Location
Chr. 18: 26133847 - 26193355 on Build GRCh38
UniGene ID
Hs.464813
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025096.1 NP_001020267.1 5 680
NM_001025097.1 NP_001020268.1 5 566
NM_001308188.1 NP_001295117.1 6 750
NM_144662.2 NP_653263.2 5 698
XM_011525818.1 XP_011524120.1 6 633
GenBank mRNA BC021713.1 - 6 690
BC025389.1 - 5 676
BC025393.1 - 5 554
BC028371.1 - 5 554 4828792
BC028686.1 - 5 554 4828931
BC042820.1 - 5 680 5268996
BC047355.1 - 5 672
BI461560.1 - 5 687

View Full Product Details