Gene Transcripts

Gene Symbol
TMEM86A
Entrez Gene ID
144110
Gene Name
transmembrane protein 86A
Gene Aliases
Chromosome Location
Chr. 11: 18698735 - 18704785 on Build GRCh38
UniGene ID
Hs.502100
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153347.2 NP_699178.1 3 704
GenBank mRNA AK074600.1 - 3 660
BC017073.2 - 2 492
BC035692.1 - 3 635 5759114

View Full Product Details