Gene Transcripts

Gene Symbol
BLOC1S3
Entrez Gene ID
388552
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 3
Gene Aliases
BLOS3, HPS8, RP
Chromosome Location
Chr. 19: 45178745 - 45202715 on Build GRCh38
UniGene ID
Hs.103902
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212550.4 NP_997715.1 1 43
GenBank mRNA AK296695.1 - 1 41
DC422050.1 - 1 43
DN996082.1 - 1 23

View Full Product Details