Gene Transcripts

Gene Symbol
IL4I1
Entrez Gene ID
259307
Gene Name
interleukin 4 induced 1
Gene Aliases
FIG1, LAAO, LAO
Chromosome Location
Chr. 19: 49889654 - 49929539 on Build GRCh38
UniGene ID
Hs.574492
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258017.1 NP_001244946.1 8, 9 1236
NM_001258018.1 NP_001244947.1 8, 9 1318
NM_152899.1 NP_690863.1 6, 7 709
NM_172374.2 NP_758962.1 8, 9 1260
NR_047577.1 9, 10 1295
GenBank mRNA AF293462.1 - 6, 7 709
AJ880386.1 - 8, 9 1135
AJ880387.1 - 8, 9 1112
AJ880388.1 - 8, 9 1174
AJ880389.1 - 9, 10 1172
AY358933.1 - 6, 7 709
BC026103.1 - 8, 9 1164
BC064378.1 - 6, 7 695 5725335
BC090852.1 - 6, 7 696 6191892
BC131625.1 - 8, 9 1149 40117891
DQ079587.1 - 8, 9 1260
DQ079588.1 - 8, 9 1318
DQ079589.1 - 8, 9 1236

View Full Product Details