Gene Transcripts

Gene Symbol
ZNF678
Entrez Gene ID
339500
Gene Name
zinc finger protein 678
Gene Aliases
Chromosome Location
Chr. 1: 227563519 - 227677443 on Build GRCh38
UniGene ID
Hs.30323
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178549.3 NP_848644.2 4 590
NR_033184.1 4 618
GenBank mRNA BC042500.1 - 4 594
CD655825.1 - 4 525

View Full Product Details