Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000318.2 | NP_000309.1 | 4 | 825 | |
NM_001079867.1 | NP_001073336.1 | 3 | 793 | ||
NM_001172086.1 | NP_001165557.1 | 5 | 831 | ||
NM_001172087.1 | NP_001165558.1 | 3 | 689 | ||
GenBank mRNA | AK312997.1 | - | 2 | 492 | |
BC000661.2 | - | 4 | 598 | 3347824 | |
BC005375.1 | - | 3 | 594 | 3934225 | |
BC093043.1 | - | 4 | 611 | 30399827 | |
M85038.1 | - | 4 | 632 | ||
M86852.1 | - | 4 | 740 |