Gene Transcripts

Gene Symbol
GYS1
Entrez Gene ID
2997
Gene Name
glycogen synthase 1
Gene Aliases
GSY, GYS
Chromosome Location
Chr. 19: 48968125 - 48993353 on Build GRCh38
UniGene ID
Hs.386225
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161587.1 NP_001155059.1 10, 11 1472
NM_002103.4 NP_002094.2 11, 12 1664
NR_027763.1 10, 11 1482
GenBank mRNA AK223579.1 - 11, 12 1620
AK291672.1 - 11, 12 1620
AK294349.1 - 6, 7 580
AK300369.1 - 11, 12 1452
AK302738.1 - 10, 11 1438
BC002617.2 - 11, 12 1603 3143019
BC003182.1 - 10, 11 1429 2958297
BC007688.2 - 11, 12 1590 3953692
J04501.1 - 11, 12 1583
U32573.1 - 11, 12 1562

View Full Product Details