Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289000.1 | NP_001275929.1 | 2, 3 | 261 | |
NM_001289001.1 | NP_001275930.1 | 2, 3 | 271 | ||
NM_153609.3 | NP_705837.1 | 2, 3 | 340 | ||
XM_011529989.2 | XP_011528291.1 | 2, 3 | 399 | ||
GenBank mRNA | AJ319876.1 | - | 1, 2 | 203 | |
AY055384.1 | - | 2, 3 | 340 | ||
AY358398.1 | - | 2, 3 | 271 | ||
BC039082.1 | - | 2, 3 | 343 | 5164881 | |
CR456446.1 | - | 2, 3 | 261 |