Gene Transcripts

Gene Symbol
NDP
Entrez Gene ID
4693
Gene Name
NDP, norrin cystine knot growth factor
Gene Aliases
EVR2, FEVR, ND
Chromosome Location
Chr. X: 43948776 - 43973675 on Build GRCh38
UniGene ID
Hs.522615
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000266.3 NP_000257.1 2 663
GenBank mRNA AK313409.1 - 2 442
BC029901.1 - 1 100 5179578
X65724.1 - 2 500
X65882.1 - 2 492

View Full Product Details