Gene Transcripts
- Gene Symbol
- HTATSF1P2
- Entrez Gene ID
- 401233
- Gene Name
- HIV-1 Tat specific factor 1 pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 6: 3020156 - 3024771 on Build GRCh38
- UniGene ID
- Hs.441039
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033884.1 |
|
1 |
1540 |
|
GenBank mRNA |
AK128391.1 |
- |
1 |
665 |
|
BX648511.1 |
- |
1 |
660 |
|
View Full Product Details