Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000791.3 | NP_000782.1 | 2, 3 | 628 | |
NM_001290357.1 | NP_001277286.1 | 2, 3 | 628 | ||
GenBank mRNA | AK297302.1 | - | 2, 3 | 199 | |
AK312642.1 | - | 1 | 172 | ||
BC000192.1 | - | 2, 3 | 191 | 3352539 | |
BC003584.1 | - | 3, 4 | 590 | 3354190 | |
BC070280.1 | - | 2, 3 | 173 | 6714220 | |
BC071996.1 | - | 3, 4 | 590 | 4645596 | |
CB988944.1 | - | 2, 3 | 657 | ||
EU145592.1 | - | 1 | 136 | ||
J00140.1 | - | 2, 3 | 178 | ||
V00507.1 | - | 1 | 136 |