Gene Transcripts

Gene Symbol
RNF208
Entrez Gene ID
727800
Gene Name
ring finger protein 208
Gene Aliases
Chromosome Location
Chr. 9: 137220247 - 137221323 on Build GRCh38
UniGene ID
Hs.512767
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031297.4 NP_112587.2 1 463
GenBank mRNA AF416715.1 - 1 144
AL136540.1 - 1 146
BC016958.1 - 1 152 4151712
DN995061.1 - 1 226

View Full Product Details