Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000171.3 | NP_000162.2 | 9 | 1377 | |
NM_001146040.1 | NP_001139512.1 | 9 | 1401 | ||
NM_001292000.1 | NP_001278929.1 | 8 | 1249 | ||
XM_017009349.1 | XP_016864838.1 | 9 | 1138 | ||
GenBank mRNA | AK226046.1 | - | 8 | 1249 | |
AK312702.1 | - | 9 | 1303 | ||
BC074980.2 | - | 9 | 1131 | 30915298 | |
BC114947.1 | - | 9 | 1401 | 40084102 | |
BC114967.1 | - | 8 | 1249 | 40084103 | |
X52009.1 | - | 9 | 1386 |