Gene Transcripts

Gene Symbol
Adrb1
Entrez Gene ID
11554
Gene Name
adrenergic receptor, beta 1
Gene Aliases
Adrb-1, beta-AR
Chromosome Location
Chr. 19: 56722372 - 56724862 on Build GRCm38
UniGene ID
Mm.46797
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007419.2 NP_031445.2 1 401
GenBank mRNA AK018378.1 - 1 219
BC140970.1 - 1 574 9055582
BC147435.1 - 1 574 9056029

View Full Product Details