Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007740.3 | NP_031766.3 | 2 | 312 | |
GenBank mRNA | AK031071.1 | - | 2 | 222 | |
BC046970.1 | - | 2 | 200 | 6314986 | |
CX207941.1 | - | 2 | 312 | ||
D17511.1 | - | 2 | 211 | ||
L12215.1 | - | 1 | 67 |