Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010331.2 | NP_034461.1 | 5, 6 | 706 | |
GenBank mRNA | AB002136.1 | - | 5, 6 | 635 | |
AB006970.1 | - | 5, 6 | 642 | ||
AK077518.1 | - | 5, 6 | 706 | ||
AK164219.1 | - | 5, 6 | 652 | ||
AK164978.1 | - | 5, 6 | 706 | ||
AK168057.1 | - | 5, 6 | 909 | ||
AK192435.1 | - | 2, 3 | 203 | ||
BC006697.1 | - | 5, 6 | 645 | 3498573 |