Gene Transcripts

Gene Symbol
HOXD3
Entrez Gene ID
3232
Gene Name
homeobox D3
Gene Aliases
HOX1D, HOX4, HOX4A, Hox-4.1
Chromosome Location
Chr. 2: 176136921 - 176173102 on Build GRCh38
UniGene ID
Hs.93574
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006898.4 NP_008829.3 2 235
XM_005246509.3 XP_005246566.1 3 502
XM_005246510.4 XP_005246567.1 3 318
XM_005246511.3 XP_005246568.1 3 926
XM_005246513.4 XP_005246570.1 3 529
XM_006712477.2 XP_006712540.1 2 212
XM_011511065.2 XP_011509367.1 3 2306
XM_011511066.2 XP_011509368.1 3 380
GenBank mRNA BC005124.2 - 2 236
BC008789.2 - 1 167 3628279

View Full Product Details