Gene Transcripts

Gene Symbol
Ush1g
Entrez Gene ID
16470
Gene Name
Usher syndrome 1G
Gene Aliases
Sans, js
Chromosome Location
Chr. 11: 115315192 - 115322142 on Build GRCm38
UniGene ID
Mm.451539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176847.3 NP_789817.1 3 1925
GenBank mRNA AB087502.1 - 3 1925

View Full Product Details