Gene Transcripts

Gene Symbol
Kcnj9
Entrez Gene ID
16524
Gene Name
potassium inwardly-rectifying channel, subfamily J, member 9
Gene Aliases
1700085N21Rik, Girk3, Kir3.3, mbGIRK3
Chromosome Location
Chr. 1: 172320501 - 172329331 on Build GRCm38
UniGene ID
Mm.261168
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008429.2 NP_032455.2 4 1297
GenBank mRNA AK033970.1 - 4 1297
AK046084.1 - 4 1336
BC065161.1 - 3 1098 5718431

View Full Product Details