Gene Transcripts

Gene Symbol
L1cam
Entrez Gene ID
16728
Gene Name
L1 cell adhesion molecule
Gene Aliases
CD171, L1, N-CAM-L1, NCAM-L1
Chromosome Location
Chr. X: 73853780 - 73880841 on Build GRCm38
UniGene ID
Mm.260568
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008478.3 NP_032504.3 7 908
GenBank mRNA AK139407.1 - 7 870
BC056988.1 - 7 833 6822217
X12875.1 - 6 655

View Full Product Details