Gene Transcripts
- Gene Symbol
- Laptm4a
- Entrez Gene ID
- 17775
- Gene Name
- lysosomal-associated protein transmembrane 4A
- Gene Aliases
- AA286466, LAPTM4, MTP, Mtrp, mKIAA0108
- Chromosome Location
- Chr. 12: 8921307 - 8938742 on Build GRCm38
- UniGene ID
- Mm.30071
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008640.2 |
NP_032666.2
|
6, 7 |
1501 |
|
GenBank mRNA |
AK013963.1 |
- |
5, 6 |
519 |
|
AK075699.1 |
- |
6, 7 |
736 |
|
AK084515.1 |
- |
6, 7 |
1144 |
|
AK088958.1 |
- |
6, 7 |
761 |
|
AK165638.1 |
- |
6, 7 |
770 |
|
AK182919.1 |
- |
3, 4 |
282 |
|
BC086667.1 |
- |
6, 7 |
681 |
30633914 |
BC089021.1 |
- |
6, 7 |
714 |
6734038 |
U34259.1 |
- |
7, 8 |
1368 |
|
View Full Product Details