Gene Transcripts

Gene Symbol
Lamc3
Entrez Gene ID
23928
Gene Name
laminin gamma 3
Gene Aliases
AI562206, AW240805
Chromosome Location
Chr. 2: 31887281 - 31946535 on Build GRCm38
UniGene ID
Mm.302362
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011836.3 NP_035966.2 2 476
GenBank mRNA AF079520.1 - 2 466
AF083372.1 - 2 413
BC096366.1 - 2 476 2865309

View Full Product Details