Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000891.2 | NP_000882.1 | 2 | 1324 | |
GenBank mRNA | AF011904.1 | - | 1 | 941 | |
AF021139.1 | - | 1 | 941 | ||
AF153820.1 | - | 2 | 1324 | ||
AK314081.1 | - | 2 | 1342 | ||
U12507.1 | - | 2 | 1266 | ||
U16861.1 | - | 2 | 1268 | ||
U22413.1 | - | 1 | 941 | ||
U24055.1 | - | 1 | 1054 |