Gene Transcripts

Gene Symbol
Abcf3
Entrez Gene ID
27406
Gene Name
ATP-binding cassette, sub-family F (GCN20), member 3
Gene Aliases
AI326318, AU016058, BB119416
Chromosome Location
Chr. 16: 20548556 - 20561603 on Build GRCm38
UniGene ID
Mm.27135
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013852.2 NP_038880.1 7 840
GenBank mRNA AK030626.1 - 7 805
AK179781.1 - 1 70
BC032923.1 - 7 781 1430389

View Full Product Details