Gene Transcripts

Gene Symbol
Zdhhc8
Entrez Gene ID
27801
Gene Name
zinc finger, DHHC domain containing 8
Gene Aliases
D16H22S1738E, DHHC-8, E330009O14Rik, Op53c05, ZDHHCL1
Chromosome Location
Chr. 16: 18220748 - 18235136 on Build GRCm38
UniGene ID
Mm.326263
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172151.4 NP_742163.4 7 979
GenBank mRNA AY668947.1 - 7 853
AY894892.1 - 7 979

View Full Product Details