Gene Transcripts

Gene Symbol
Capn15
Entrez Gene ID
50817
Gene Name
calpain 15
Gene Aliases
Solh
Chromosome Location
Chr. 17: 25958364 - 25985791 on Build GRCm38
UniGene ID
Mm.216072
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015830.1 NP_056645.1 6 2360
GenBank mRNA AK079422.1 - 5 2452
AK143711.1 - 8 2999
AK148209.1 - 4 619
AK170543.1 - 7 2594
BC058094.1 - 7 2618 6824892

View Full Product Details