Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081475.1 | NP_001074944.1 | 10 | 987 | |
NM_001284229.1 | NP_001271158.1 | 9 | 906 | ||
NM_016777.3 | NP_058057.3 | 12 | 1956 | ||
GenBank mRNA | AF034610.2 | - | 12 | 1946 | |
AF095722.1 | - | 9 | 887 | ||
AK083333.1 | - | 10 | 988 | ||
BC004693.1 | - | 10 | 954 | 3498591 |