Gene Transcripts
- Gene Symbol
- Ccnc
- Entrez Gene ID
- 51813
- Gene Name
- cyclin C
- Gene Aliases
- AI451004, AU020987, CG1C
- Chromosome Location
- Chr. 4: 21727701 - 21759922 on Build GRCm38
- UniGene ID
- Mm.278584
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001122982.2 |
NP_001116454.1
|
5, 6 |
523 |
|
NM_001290420.1 |
NP_001277349.1
|
5, 6 |
523 |
|
NM_001290422.1 |
NP_001277351.1
|
5, 6 |
523 |
|
NM_016746.4 |
NP_058026.2
|
5, 6 |
523 |
|
GenBank mRNA |
AK009615.1 |
- |
5, 6 |
468 |
|
AK014079.1 |
- |
4, 5 |
372 |
|
AK037957.1 |
- |
5, 6 |
524 |
|
AK135472.1 |
- |
5, 6 |
519 |
|
AK153035.1 |
- |
5, 6 |
456 |
|
BB631264.1 |
- |
5, 6 |
521 |
|
BC003344.1 |
- |
5, 6 |
456 |
3257730 |
BC037689.1 |
- |
5, 6 |
451 |
|
BC062376.1 |
- |
5, 6 |
438 |
|
BC087544.1 |
- |
5, 6 |
455 |
|
BC100396.1 |
- |
5, 6 |
497 |
|
BC120649.1 |
- |
5, 6 |
450 |
40129572 |
BC120677.1 |
- |
5, 6 |
450 |
40129600 |
U62638.1 |
- |
5, 6 |
498 |
|
View Full Product Details